missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ 20
GREENER_CHOICE

Product Code. p-5545700
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18780109

Brand: Tocris Bioscience™ 5198/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ATR kinase inhibitor; antitumor

AZ 20 is a potent and selective ATR kinase inhibitor (IC50 = 5 nM). Exhibits 7.6-fold selectivity over mTOR and selectivity over a panel of 442 kinases, including ATM kinase, PI3-K isoforms, and DNA-PK. Inhibits cell growth in cell lines with high baseline levels of replication stress. Displays antitumor effects in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type AZ 20
Purity >98%
Molecular Weight (g/mol) 412.51
Inhibitors AZ 20

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.