missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ1 USP25/28 inhibitor
GREENER_CHOICE

Product Code. 18725905
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18725905

Brand: Tocris Bioscience™ 7845/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective USP25 and USP28 inhibitor

AZ1 USP25/28 inhibitor is a selective inhibitor of USP28 (IC50 = 0.6 μM, Kd= 0.2 - 3.7 μM) and USP25 (IC50 = 0.7 μM). AZ1 USP25/28 inhibitor exhibits selectivity for USP25 and USP28 over other DUB family members. The compound dose-dependently reduces c-Myc levels in colon carcinoma cells in vitro and induces cell death (EC50 values are 18 - 20 μM). In a mouse Alzheimer's disease model, AZ1 USP25/28 inhibitor reduces amyloid burden, attenuates microglial activation and improves synaptic and cognitive function.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[[5-Bromo-2-[[4-fluoro-3-(trifluoromethyl)phenyl]methoxy]phenyl]methyl]amino]ethanol
CAS 2165322-94-9
Quantity 10 mg
Target USP28
Molecular Formula C17H16BrF4NO2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.