missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ1 USP25/28 inhibitor

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AZ1 USP25/28 inhibitor is a selective inhibitor of USP28 (IC50 = 0.6 μM, Kd= 0.2 - 3.7 μM) and USP25 (IC50 = 0.7 μM). AZ1 USP25/28 inhibitor exhibits selectivity for USP25 and USP28 over other DUB family members. The compound dose-dependently reduces c-Myc levels in colon carcinoma cells in vitro and induces cell death (EC50 values are 18 - 20 μM). In a mouse Alzheimer's disease model, AZ1 USP25/28 inhibitor reduces amyloid burden, attenuates microglial activation and improves synaptic and cognitive function.
Specifications
Specifications
| Chemical Name or Material | 2-[[[5-Bromo-2-[[4-fluoro-3-(trifluoromethyl)phenyl]methoxy]phenyl]methyl]amino]ethanol |
| CAS | 2165322-94-9 |
| Quantity | 50 mg |
| Target | USP28 |
| Molecular Formula | C17H16BrF4NO2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction