missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-PEG4-alcohol

Product Code. 15436489
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436489 100 mg 100mg
15426489 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436489 Supplier Thermo Scientific Alfa Aesar Supplier No. J64617.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ADIBO-containing reagent with PEG4 spacer and alcohol terminus | C29H36N2O7

ADIBO-containing reagent with PEG4 spacer and alcohol terminus

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C29H36N2O7
MDL Number MFCD22380739
Synonym ADIBO-PEG4-alcohol

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 524.61
Physical Form Viscous Liquid or Solid
Chemical Name or Material Azadibenzocyclooctyne-PEG4-alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.