missing translation for 'onlineSavingsMsg'
Learn More

Azadirachtin D, TRC

Product Code. 30012328
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012328 1 mg 1mg
30009867 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012328 Supplier Toronto Research Chemicals Supplier No. TRCA7973301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 99399-65-2
Molecular Formula C34H44O14
Molecular Weight (g/mol) 676.7
Synonym 1H,7H-Naphtho[1,8-bc:4,4a-c']difuran-5-carboxylic acid, 10-(acetyloxy)decahydro-3,5-dihydroxy-4,10a-dimethyl-8-[[(2E)-2-methyl-1-oxo-2-buten-1-yl]oxy]-4-[(1aR,2S,3aS,6aS,7S,7aS)-3a,6a,7,7a-tetrahydro-6a-hydroxy-7a-methyl-2,7-methanofuro[2,3-b]oxireno[e]oxepin-1a(2H)-yl]-, methyl ester, (2aS,3S,4S,4aR,5S,7aS,8S,10R,10aR,10bS)-, 1H,7H-Naphtho[1,8-bc:4,4a-c']difuran-5-carboxylic acid, 10-(acetyloxy)decahydro-3,5-dihydroxy-4,10a-dimethyl-8-[[(2E)-2-methyl-1-oxo-2-butenyl]oxy]-4-[(1aR,2S,3aS,6aS,7S,7aS)-3a,6a,7,7a-tetrahydro-6a-hydroxy-7a-methyl-2,7-methanofuro[2,3-b]oxireno[e]oxepin-1a(2H)-yl]-, methyl ester, (2aS,3S,4S,4aR,5S,7aS,8S,10R,10aR,10bS)- (9CI), Methyl (2aS,3S,4S,4aR,5S,7aS,8S,10R,10aR,10bS)-10-(acetyloxy)decahydro-3,5-dihydroxy-4,10a-dimethyl-8-[[(2E)-2-methyl-1-oxo-2-buten-1-yl]oxy]-4-[(1aR,2S,3aS,6aS,7S,7aS)-3a,6a,7,7a-tetrahydro-6a-hydroxy-7a-methyl-2,7-methanofuro[2,3-b]oxireno[e]oxepin-1a(2H)-yl]-1H,7H-naphtho[1,8-bc:4,4a-c']difuran-5-carboxylate, Azadirachtin D
IUPAC Name methyl (1S,4S,5R,6S,7S,8S,11R,12R,14S,15S)-12-acetyloxy-4,7-dihydroxy-6-[(1S,2S,6S,8S,9R,11S)-2-hydroxy-11-methyl-5,7,10-trioxatetracyclo[6.3.1.02,6.09,11]dodec-3-en-9-yl]-6,11-dimethyl-14-[(E)-2-methylbut-2-enoyl]oxy-3,9-dioxatetracyclo[6.6.1.01,5.011,15]pentadecane-4-carboxylate
SMILES O[C@]1(C=CO2)[C@]2([H])O[C@]3([H])[C@]4([C@]5([C@@]([C@@]6(O)C(OC)=O)([H])[C@]([C@H](C[C@H]7OC(C)=O)OC(/C(C)=C/C)=O)(CO6)[C@@]8([H])[C@@](OC[C@@]87C)([H])[C@H]5O)C)[C@@](C)(O4)[C@@]1([H])C3

Specifikationer

Quantity 1 mg
InChI Formula InChI=1S/C34H44O14/c1-8-15(2)24(37)46-19-12-18(45-16(3)35)28(4)13-43-21-22(28)31(19)14-44-33(40,26(38)41-7)25(31)29(5,23(21)36)34-20-11-17(30(34,6)48-34)32(39)9-10-42-27(32)47-20/h8-10,17-23,25,27,36,39-40H,11-14H2,1-7H3/b15-8+/t17-,18-,19+,20+,21+,22+,23-,25+,27+,28-,29-,30+,31+,32+,33+,34+/m1/s1
Chemical Name or Material Azadirachtin D
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.