missing translation for 'onlineSavingsMsg'
Learn More

Azaperone

Product Code. 15597529 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15597529 1 g 1g
15507539 5 g 5g
15517539 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15597529 Supplier Thermo Scientific Acros Supplier No. 460830010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1649-18-9 | C19H22FN3O | 327.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1649-18-9
Molecular Formula C19H22FN3O
Molecular Weight (g/mol) 327.40
MDL Number MFCD00866692
InChI Key XTKDAFGWCDAMPY-UHFFFAOYSA-N
Synonym azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum
PubChem CID 15443
IUPAC Name 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
SMILES FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1

Specifications

Loss on Drying 0.5% max. (60°C, 4 h) (vacuum)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, chloroform
Formula Weight 327.4
Chemical Name or Material Azaperone
compliance-icons
Product Identifier
  • Azaperone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.