missing translation for 'onlineSavingsMsg'
Learn More

Azatadine Dimaleate, TRC

Product Code. 30180216
Change view
Click to view available options
Quantity:
5 mg
50 mg
250 mg
Unit Size:
250mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180216 5 mg 5mg
30180025 50 mg 50mg
30179842 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180216 Supplier Toronto Research Chemicals Supplier No. TRCA8029505MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3978-86-7
Molecular Formula C20 H22 N2 . 2 C4 H4 O4
Molecular Weight (g/mol) 522.55
Synonym 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 6,11-dihydro-11-(1-methyl-4-piperidinylidene)-, (2Z)-2-butenedioate (1:2), 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 6,11-dihydro-11-(1-methyl-4-piperidinylidene)-, (Z)-2-butenedioate (1:2), 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 6,11-dihydro-11-(1-methyl-4-piperidylidene)-, maleate (1:2) (8CI), 5-(4'-N-Methylpiperidylidene)-4-azo-10:11-dihydrodibenzocycloheptene dimaleate, Atoramin, Azatadine dimaleate, Azatadine maleate, Bonamid, Idulian, Optimine, Sch 10649, Trinalin, Zadine
IUPAC Name (Z)-but-2-enedioic acid;11-(1-methylpiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[3,4-b]pyridine
SMILES CN1CCC(=C2c3ccccc3CCc4cccnc24)CC1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H22N2.2C4H4O4/c1-22-13-10-16(11-14-22)19-18-7-3-2-5-15(18)8-9-17-6-4-12-21-20(17)19;2*5-3(6)1-2-4(7)8/h2-7,12H,8-11,13-14H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formula Weight 522.2002
Chemical Name or Material Azatadine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.