missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AZD 4547
Description
AZD 4547 is a potent and selective FGFR1, FGFR3 and FGFR2 tyrosine kinase inhibitor (IC50 values are 0.2 1.8, and 2.5 nM, respectively). It shows selectivity for FGFR across a range of unrelated tyrosine and serine/threonine kinases, including IGFR (>2,900-fold), CDK2 (>50,000-fold), and p38 (>50,000-fold). In vitro, it has antiproliferative effects on tumor cell lines with deregulated FGFR expression. In vivo, AZD 4547 exhibits dose-dependent inhibition of tumor growth in an FGFR-driven human tumor xenograft mice model. It suppresses osteoclastogenesis and tumor-induced osteolysis in an orthotopic breast cancer bone metastasis mouse model. AZD 4547 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | rel-N-[5-[2-(3,5-Dimethoxyphenyl)ethyl]-1H-pyrazol-3-yl]-4-[(3R,5S)-3,5-dimethyl-1-piperazinyl]benzamide |
| CAS | 1035270-39-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | FGF Receptor Inhibitors |
| Molecular Formula | C26H33N5O3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?