missing translation for 'onlineSavingsMsg'
Learn More

Azelaic acid, 85-90%, tech., flakes, ACROS Organics™

Product Code. 10214610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10214610 25g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10214610 Supplier Thermo Scientific Acros Supplier No. 105120250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 123-99-9
Color White
Assay Percent Range 85 to 90%
Molecular Formula C9H16O4
Linear Formula HOOC(CH2)7COOH
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILES C(CCCC(=O)O)CCCC(=O)O
IUPAC Name nonanedioic acid
Molecular Weight (g/mol) 188.223
PubChem CID 2266
ChEBI CHEBI:48131
Formula Weight 188.22
Percent Purity ≥85%
Grade Technical
Quantity 25 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Flakes
Chemical Name or Material Azelaic acid, tech., 85-90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.