missing translation for 'onlineSavingsMsg'
Learn More

Azelaic acid, 98%

Product Code. 13469508 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13469508 500 g Glass Bottle 500g
10476832 25 g Glass Bottle 25g
10477022 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 13469508 Fornitore Thermo Scientific Acros N. del fornitore 401525000
 Restricted Item
Estimated Shipment: 06-11-2026
Aggiungi al carrello
Edge
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifica

Melting Point 98.0°C to 103.0°C
Boiling Point 237.0°C (20.0hPa)
Color White to Yellow
Quantity 500 g
Flash Point 215°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Slightly Crystalline Powder or Flakes
Assay Percent Range 98%
Linear Formula HOOC(CH2)7COOH
Solubility Information Solubility in water: 2.4g/L water (20°C). Other solubilities: very soluble in alcohol and ether:2.7g/100mL(15°C), soluble in organic solvents, slightly soluble in benzene
Formula Weight 188.22
Percent Purity 98%
Chemical Name or Material Azelaic Acid, 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.