missing translation for 'onlineSavingsMsg'
Learn More

Azelaic acid, tech. 80%

Product Code. 11483497
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483497 500 g 500g
11493497 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483497 Supplier Thermo Scientific Alfa Aesar Supplier No. A17271.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol

It is applied in organic synthesis, lacquers, polyester adhesives, and urethane elastomers. Azelaic acid directly inhibits kallikrein 5 (KLK5), down regulating expression of KLK5, toll-like receptor 2 (TLR 2), and cathelicidin, and suppresses activity of serine proteases. In PUFA-treated fibroblasts, azelaic acid decreases generation of ROS and up regulates expression of antioxidative enzymes, inhibiting cell damage; agonist activity at PPAR? is thought to drive these effects. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifications

Melting Point ∼102°C
Boiling Point 286°C (100mmHg)
Beilstein 1101094
Merck Index 14,905
Solubility Information Soluble in water at 20°C 2.4g/L. Soluble in ethanol (5%),DMSO,methanol (70mg/ml),and alcohol.
Formula Weight 188.22
Percent Purity 80%
Grade Technical
Density 1.225
Quantity 500 g
Flash Point 215°C (419°F)
Chemical Name or Material Azelaic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.