missing translation for 'onlineSavingsMsg'
Learn More

Azelaic Acid, TRC

Product Code. 17952165
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952165 25 g 25g
30134239 1 g 1g
17910465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952165 Supplier Toronto Research Chemicals Supplier No. TRCA80814025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Formula Weight 188.1049
InChI Formula InChI=1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)
IUPAC Name nonanedioic acid
Molecular Formula C9 H16 O4
Molecular Weight (g/mol) 188.22
Percent Purity >95%
SMILES OC(=O)CCCCCCCC(=O)O
Synonym Azelaic Acid, Nonanedioic acid, 1,7-Heptanedicarboxylic acid, Lepargylic acid, Anchoic acid, Azelex, Skinoren
Chemical Name or Material 1,7-Heptanedicarboxylic acid
Grade HPLC
Show More Show Less

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.