missing translation for 'onlineSavingsMsg'
Learn More

Azelastine hydrochloride, MedChemExpress

Product Code. 16428326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428326 100 mg 100mg
16421426 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428326 Supplier Medchem Express Supplier No. HYB0462
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azelastine hydrochloridem, an antihistamine, is a potent and selective histamine 1 (H1) antagonist. Azelastine hydrochloride can be used for the research of allergic rhinitis, asthma, diabetic hyperlipidemic and SARS-CoV-2.

Azelastine hydrochloridem, an antihistamine, is a potent and selective histamine 1 (H1) antagonist. Azelastine hydrochloride can be used for the research of allergic rhinitis, asthma, diabetic hyperlipidemic and SARS-CoV-2.

Specifications

Chemical Name or Material Azelastine hydrochloride
CAS 79307-93-0
Color White
Purity Grade Notes Research
Molecular Formula C22H25Cl2N3O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (119.51 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (15.94 mM; Need ultrasonic)
SMILES O=C1N(C2CCN(C)CCC2)N=C(CC3=CC=C(Cl)C=C3)C4=C1C=CC=C4.Cl
Molecular Weight (g/mol) 418.36
Formula Weight 418.36
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.