missing translation for 'onlineSavingsMsg'
Learn More

Azelastine hydrochloride

Product Code. 16564304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16564304 250 mg 250mg
16554304 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16564304 Supplier Thermo Scientific Acros Supplier No. 465082500
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79307-93-0 | C22H25Cl2N3O | 418.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79307-93-0
Molecular Formula C22H25Cl2N3O
Molecular Weight (g/mol) 418.36
MDL Number MFCD00242783
InChI Key YEJAJYAHJQIWNU-UHFFFAOYNA-N
IUPAC Name hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
SMILES [H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O

Specifications

Melting Point 225.0°C to 229.0°C
Quantity 250 mg
Formula Weight 418.36
Chemical Name or Material Azelastine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.