missing translation for 'onlineSavingsMsg'
Learn More

Azetidine hydrochloride, 97%

Product Code. 15456388
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456388 1 g 1g
15466388 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456388 Supplier Thermo Scientific Alfa Aesar Supplier No. H59935.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 36520-39-5 | C3H8ClN | 93.554 g/mol

Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

Solubility
Soluble in water (103.8°C at 760 mmHg).

Notes
Hygroscopic. Reacts with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36520-39-5
Molecular Formula C3H8ClN
Molecular Weight (g/mol) 93.554
MDL Number MFCD00191762
InChI Key HGQULGDOROIPJN-UHFFFAOYSA-N
Synonym azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
PubChem CID 12308726
IUPAC Name azetidine;hydrochloride
SMILES C1CNC1.Cl

Specifications

Melting Point >300°C
Flash Point 152°C (306°F)
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (103.8°C at 760mmHg).
Formula Weight 93.56
Percent Purity 97%
Chemical Name or Material Azetidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.