missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-alcohol, TRC

Product Code. 30347100
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347100 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347100 Supplier Toronto Research Chemicals Supplier No. TRCA8485505G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-(2-Azidoethoxy)ethoxy]ethanol
CAS 86520-52-7
Molecular Formula C6 H13 N3 O3
Quantity 5 g
Synonym 8-Azido-3,6-dioxaoctanol, 1-Azido-8-hydroxy-3,6-dioxaoctane, 2-[2-(2-Azidoethoxy)ethoxy]ethanol
InChI Formula InChI=1S/C6H13N3O3/c7-9-8-1-3-11-5-6-12-4-2-10/h10H,1-6H2
SMILES OCCOCCOCCN=[N+]=[N-]
IUPAC Name 2-[2-(2-azidoethoxy)ethoxy]ethanol
Molecular Weight (g/mol) 175.19
Formula Weight 175.0957
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.