missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-biotin conjugate

Product Code. 15496679
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496679 25 mg 25mg
15406689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496679 Supplier Thermo Scientific Alfa Aesar Supplier No. J64996.MB
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media | CAS: 875770-34-6 | C18H32N6O5S | 444.551 g/mol

Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it?s components do not react with the biological environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Solubility
Solubile in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875770-34-6
Molecular Formula C18H32N6O5S
Molecular Weight (g/mol) 444.551
MDL Number MFCD20134145
InChI Key ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
Synonym biotin-peg3-azide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl biotinamide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamide, biotin-peg4-n3, azide-peg3-biotin conjugate, azido-peg3-biotin conjugate, 11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane, 3abeta,6abeta-n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-2-oxohexahydro-1h-thieno 3,4-d imidazole-4alpha-pentanamide
PubChem CID 60146223
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2

Specifications

Color White to Gray
Quantity 25 mg
Solubility Information Solubile in DMSO,DMF and water.
Formula Weight 444.55
Physical Form Solid
Chemical Name or Material Azido-PEG3-biotin conjugate
compliance-icons
Product Identifier
  • Azido-PEG3-biotin conjugate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Azido-PEG4-biotin conjugate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.