missing translation for 'onlineSavingsMsg'
Learn More

Azidoethanol, TRC

Product Code. 30136594
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136594 5 g 5g
30136239 10 g 10g
30004836 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136594 Supplier Toronto Research Chemicals Supplier No. TRCA8485605G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-05-1
Molecular Formula C2 H5 N3 O
Molecular Weight (g/mol) 87.08
Synonym Ethanol, 2-azido-, -Azido-2-hydroxyethane, 2-Azido-1-ethanol, 2-Azidoethanol, 2-Hydroxyethyl azide, Azidoethanol, 2-Azidoethanol
IUPAC Name 2-azidoethanol
SMILES OCCN=[N+]=[N-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H5N3O/c3-5-4-1-2-6/h6H,1-2H2
Formula Weight 87.0433
Percent Purity >95%
Chemical Name or Material 2-Azidoethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.