Learn More
Tocris Bioscience™ Azidothymidine
Selective reverse transcriptase inhibitor; antiretroviral
Brand:  Tocris Bioscience™ 4150/50
Encompass
Product Code. 15716555
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Selective reverse transcriptase inhibitor. Exhibits 100-fold selectivity for HIV reverse transcriptase over DNA polymerase α. Suppresses HIV-1 replication and enhances cell viability in HIV-1 infected T cells. Attenuates growth of multiple myeloma (MM) cells in vitro and reduces growth of MM tumor xenografts in mice. Increases differentiation iPSCs into cortical neurons and reduces the over-proliferation of neural precursors. Orally bioavailable and brain penetrant. Antiretroviral agent. Also decreases CRISPR-mediated homology directed repair (HDR) efficiency.
Specifications
Specifications
99%
30516-87-1
50 mg
3 '-Azido-3 '-deoxythymidine
-20°C