missing translation for 'onlineSavingsMsg'
Learn More

Azidotrimethylsilane, 94%

Product Code. 11985801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
Product Code. Quantity Packaging unitSize
11985801 250 mL Glass bottle 250mL
10429270 10 mL Glass bottle 10mL
10763551 50 mL Glass bottle 50mL
3 options
This item is not returnable. View return policy

Product Code. 11985801

Brand: Thermo Scientific Acros 167932500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
IUPAC Name azido(trimethyl)silane
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Color Colorless to Yellow
Density 0.8760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiN3
Refractive Index 1.4142 to 1.4162
Quantity 250 mL
Fieser 01,1236; 03,316; 05,719; 06,632; 07,394; 09,21; 10,14; 11,32; 12,37; 13,24; 14,25; 15,16; 16,17; 17,378
Specific Gravity 0.876
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, dichloromethane and benzene
Formula Weight 115.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Azidotrimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Azidotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.