missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan Medoxomil, TRC

Product Code. 30133130
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133130 500 mg 500mg
30008741 100 mg 100mg
30012697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133130 Supplier Toronto Research Chemicals Supplier No. TRCA926910500MG
 Restricted Item
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863031-21-4
Molecular Formula C30 H24 N4 O8
Molecular Weight (g/mol) 568.53
Synonym 1H-Benzimidazole-7-carboxylic acid, 1-[[2'-(2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-1-[[2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Azilsartan Medoxomil, TAK 491
IUPAC Name (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-3-[[4-[2-(5-oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMILES CCOc1nc2cccc(C(=O)OCC3=C(C)OC(=O)O3)c2n1Cc4ccc(cc4)c5ccccc5C6=NOC(=O)N6

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C30H24N4O8/c1-3-38-28-31-23-10-6-9-22(27(35)39-16-24-17(2)40-30(37)41-24)25(23)34(28)15-18-11-13-19(14-12-18)20-7-4-5-8-21(20)26-32-29(36)42-33-26/h4-14H,3,15-16H2,1-2H3,(H,32,33,36)
Formula Weight 568.1594
Percent Purity >95%
Chemical Name or Material Azilsartan Medoxomil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.