missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE DIY Printed Safety Wash Bottles

Product Code. 12590056
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Type:
Non-venting Cap
Vapor-venting
Closure Color:
Assorted
White
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Type Closure Color unitSize
12590056 500 mL Vapor-venting White Pack of 5
12540066 250 mL Vapor-venting White Pack of 5
12510066 250 mL Non-venting Cap White Pack of 5
12520066 500 mL Non-venting Cap White Pack of 5
15307047 500 mL Vapor-venting Assorted Each
12500066 1000 mL Vapor-venting White Pack of 5
12530066 1000 mL Non-venting Cap White Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12590056 Supplier Azlon™ Supplier No. WGW002VTML
Only null left
Add to basket
Edge
Add to basket

Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.

  • Do-it-yourself wash bottles with blank write-on panel
  • Indelibly printed in three languages (English, French and German)
  • Supplied with adhesive hazard labels
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.91 oz.
Color Translucent
Diameter (Metric) Outer Neck 38 mm
Diameter (Metric) Inner Neck 34 mm
Includes Closure, blank write-on panel, adhesive hazard signs
Material LDPE
Quantity 5/PK
Temperature Range (Metric) –50 to 80°C
Closure Color White
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 163 mm
Product Type DIY Printed Safety Wash Bottle
Closure Material Polypropylene
Diameter (Metric) 60 mm
Closure Type Vapor-venting
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.