missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ LDPE DIY Printed Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Type:
Non-venting Cap
Vapor-venting
Closure Color:
Assorted
White
Unit Size:
Each
Pack of 5
Description
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
- Do-it-yourself wash bottles with blank write-on panel
- Indelibly printed in three languages (English, French and German)
- Supplied with adhesive hazard labels
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Diameter (Metric) Outer Neck | 38 mm |
| Diameter (Metric) Inner Neck | 34 mm |
| Includes | Closure, blank write-on panel, adhesive hazard signs |
| Material | LDPE |
| Quantity | 5/Pk. |
| Temperature Range (Metric) | -50°C to +80°C |
| Closure Color | Assorted |
| Closure Included | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction