Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
Product Code. p-7160086
Change view
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Unit Size:
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Label Unit Size
15888467 Distilled Water Pack of 5
15898467 Acetone Pack of 5
15808477 Methanol Pack of 5
15818477 Isopropanol Pack of 5
15828477 Ethanol Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15888467 Supplier Azlon™ Supplier No. WGW801P
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Azlon™ Multi-lingual Round Integral Wash Bottle is molded with an integral spout to reduce contamination caused by the inner tube touching the benchtop during re-filling.
  • Multilingual
  • Wide neck wash bottle
  • Indelibly printed in English, French, German and Spanish
  • Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
  • Fitted with color coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
Specifications
Capacity (Metric) 500 mL
Includes Integral spout
Closure Color White
Closure Included Yes
Mouth Wide
Chemical Label Distilled Water
Closure Material Polypropylene
Product Type Wash Bottle