missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polyacetal Trough for Slide Staining Kit

Product Code. 12053344
Change view
Click to view available options
Material:
Polyacetal
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12053344 Polyacetal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12053344 Supplier Azlon Supplier No. SWM016

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Slide Staining Kit

  • Dimensions - 100 x 85 x 55mm
  • Supplied complete with a push-fit lid and small adapter to aid drainage

Specifications

Color Black
Type Staining Trough
Material Polyacetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.