Learn More
Azlon™ Polypropylene Multi-lingual Printed Wash Bottles
Product Code. 15327017
Change view
Click to view available options
Chemical Label:
Dichloromethane
Methyl Ethyl Ketone
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Label Unit Size
15327017 Methyl Ethyl Ketone Pack of 5
15317017 Dichloromethane Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15327017 Supplier Azlon™ Supplier No. WGW703PP
  • ONLINE EXCLUSIVE PRICE
    52.57 €
    39.65 € / Pack of 5
    Save 12.92 €
    24% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Imprinted with Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Azlon™ Polypropylene Multi-lingual Printed Wash Bottles display chemical names and key safety information in four European languages, eliminating the need for multiple wash bottles across European language speaking countries.
  • Multi-lingual wide neck wash bottles
  • Manufactured from polypropylene for high thermal and chemical resistance; Autoclavable
  • Indelibly printed In English, French, German and Spanish
  • Fitted with color-coded, one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric) 500 mL
Color Translucent
Material Polypropylene
Quantity 5/PK
Closure Color Green
Mouth Wide
Chemical Label Methyl Ethyl Ketone
Closure Material Polypropylene
Closure Type Non-venting Cap
Autoclavable Autoclavable