Learn More
Azlon™ Screw caps for Wide Mouth Vented Wash Bottles
Product Code. 15347057
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15347057 Red Pack of 5
15377057 Blue Pack of 5
15367057 Green Pack of 5
15387057 Orange Pack of 5
15337057 White Pack of 5
15357057 Yellow Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15347057 Supplier Azlon Supplier No. WBCAPVR
  • ONLINE EXCLUSIVE PRICE
    22.28 €
    19.82 € / Pack of 5
    Save 2.46 €
    11% Off
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Caps for wide neck wash bottles with dip-tube and patented DripLok™ vapor venting valve that prevents solvent drips.
  • Available in six colors
  • One-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Color Red
Quantity 5/Pk.
Closure Material Polypropylene