Learn More
Azlon™ Silicone Rubber Hand/Finger Protector
Product Code. 12098831
Change view
Click to view available options
Size:
Finger
Medium
Small
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12098831 Medium Each
12018841 Finger Each
12008841 Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12098831 Supplier Azlon Supplier No. PGR402
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Ideal for safe handling of hot beakers, flasks
  • Capable of withstanding temperatures up to 250°C
  • For handling cold objects down to -57°C
  • Contact surfaces are multi-studded for positive grip
Specifications
Material Silicone
Product Type Hand/Finger Protector
Size Medium
For Use With (Application) For Safe Handling Of Hot Beakers, Flasks etc