missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE Tracker Clinical Wash Bottles

Product Code. 12861464
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Closure Type:
Non-venting Cap
Vapor-venting
Closure Color:
Green
Red
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Type Closure Color Unit Size
12861464 250 mL Vapor-venting Red Pack of 5
12851464 250 mL Non-venting Cap Green Pack of 5
12871464 500 mL Non-venting Cap Red Pack of 5
10092794 500 mL Vapor-venting Red Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12861464 Supplier Azlon Supplier No. WTR250VT
  • ONLINE EXCLUSIVE PRICE
    36.25 €
    32.03 € / Pack of 5
    Save 4.22 €
    11% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed with complete labelling facility to ensure full traceability

  • Meets industry standards including those required by the Clinical Pathology Accreditation body
  • Ideal for use in clinical and general hospital market
  • Unique write-on, wipe-off panel
  • DripLok™ vapour venting cap version available - for ultimate safety with volatile solvents
  • Supplied with set of COSHH labels

Specifications

Capacity (Metric) 250 mL
Color Clear
For Use With (Application) For clinical and general hospital market
Material LDPE
Closure Color Red
Closure Included Yes
Closure Material Polypropylene
Closure Type Vapor-venting
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.