missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene, TRC

Product Code. 30340643
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340643 100 mg 100mg
30335060 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30340643 Supplier Toronto Research Chemicals Supplier No. TRCA931000100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azobenzene
CAS 103-33-3
Molecular Formula C12 H10 N2
Quantity 100 mg
Synonym Azobenzene (8CI), Benzene, azo- (3CI), Diazene, diphenyl- (9CI), 1,2-Diphenyldiazene, A 15073, Azobenzide, Azobenzol, Benzene, azobis-, Benzeneazobenzene, Diphenyldiazene, Diphenyldiimide, NSC 2102, Phenylazobenzene
InChI Formula InChI=1S/C12H10N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H/b14-13+
SMILES c1ccc(cc1)N=Nc2ccccc2
IUPAC Name (E)-diphenyldiazene
Molecular Weight (g/mol) 182.22
Formula Weight 182.0844
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.