missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azomethine H, Pure, for Colorimetric Determination Of Boron, Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| CAS | 32266-60-7 |
| Packaging | Plastic powder jar |
| MDL Number | 42050 |
| Quantity | 10 g |
| Formula Weight | 423.421g/mol |
| Physical Form | Solid |
Safety and Handling
Product Identifier
- Azomethine H
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?