missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam MP Biomedicals

Product Code. 11493271 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493271 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493271 Supplier MP Biomedicals Supplier No. 02150415.5
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Quantity 500 mg
Synonym trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid, 2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
Molecular Weight (g/mol) 435.426
PubChem CID 56842327
Formula Weight 435.4
Chemical Name or Material Aztreonam
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.