missing translation for 'onlineSavingsMsg'
Learn More

Azure A (Technical Grade), TRC

Product Code. 30136805
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136805 50 g 50g
17942915 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136805 Supplier Toronto Research Chemicals Supplier No. TRCA96530350G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-53-3
Molecular Formula C14H14N3S .Cl
Molecular Weight (g/mol) 291.8
Synonym 3-Amino-7-(dimethylamino)phenazathionium Chloride, 5-Chloro-3-dimethylamino-7-amino-5H-phenothiazine, Azur A, Azure A Chloride, Azure Blue Cert, Azurea Dye, C.I. 52005, N,N-Dimethylthionine;
IUPAC Name 7-N,7-N-dimethylphenothiazin-5-ium-3,7-diamine;chloride
SMILES [Cl-].CN(C)c1ccc2nc3ccc(N)cc3[s+]c2c1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C14H14N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8H,15H2,1-2H3;1H/q+1;/p-1
Formula Weight 291.0597
Chemical Name or Material Azure A (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.