missing translation for 'onlineSavingsMsg'
Learn More

Azure A

Product Code. 15463949
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463949 100 g 100g
15453949 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463949 Supplier Thermo Scientific Alfa Aesar Supplier No. J61346.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For staining cell granules and tissues | CAS: 531-53-3 | C14H14ClN3S | 291.797 g/mol

For staining cell granules and tissues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-53-3
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.797
MDL Number MFCD00012112
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym C.I. 52005
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]

Specifications

Melting Point 290°C (decomposition)
Beilstein 3922287
Merck Index 14,927
Formula Weight 291.8
Color Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Azure A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.