missing translation for 'onlineSavingsMsg'
Learn More

Azure B, MP Biomedicals™

Product Code. 11473281 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473281 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473281 Supplier MP Biomedicals Supplier No. 0215041705
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 531-55-5
Molecular Formula C15H16ClN3S
MDL Number MFCD00011935
Synonym C.I. 52010, N, N, N-Trimethylthionin
Quantity 5 g
InChI Key KFZNPGQYVZZSNV-UHFFFAOYSA-M
Physical Form Powder
SMILES [Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C
IUPAC Name 3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular Weight (g/mol) 305.82
Formula Weight 305.83
Chemical Name or Material Azure B, Stains and dyes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.