missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ B-27™ Electrophysiology Kit
Description
The B-27™ Electrophysiology Kit contains Neurobasal™ Medium, Electro and B-27™ Supplement, Electro, both specifically developed to enhance network spike rates in cultured neurons when compared with other media and supplement combinations. Electrophysiology is the fundamental technique to assess such electrical activities to study the functions and dysfunctions in cultured neurons.
Using the B-27™ Electrophysiology Kit in your neuronal studies offers:
Increased synapse densities
More spontaneous spike rates
Improved multielectrode array efficiency
More Synapses, More Signal, Simpler Testing
The B-27™ Electrophysiology Kit is an improvement to Neurobasal™ and B-27™ combination for cultured networks. Hippocampal and cortical neurons cultured in this media system show an increased density of synapses and neurotransmitter receptors which produce higher spike rates in neural networks, helping to simplifying your study.
Relevant Results for Your Electrophysiology Studies
Using Invitrogen™ B-27™ Electrophysiology Kit, allows you to get more relevant results when studying neuron physiology and the biochemical basis of normal and disease processes. The B-27™ Electrophysiology Kit can yield better results in other electrophysiological studies, including neurons used in multielectrode arrays in neurotoxicology and neuropharmacology studies.
For other applications, we offer alternative formulations of B-27™ and Neurobasal™.
Using the B-27™ Electrophysiology Kit in your neuronal studies offers:
More Synapses, More Signal, Simpler Testing
The B-27™ Electrophysiology Kit is an improvement to Neurobasal™ and B-27™ combination for cultured networks. Hippocampal and cortical neurons cultured in this media system show an increased density of synapses and neurotransmitter receptors which produce higher spike rates in neural networks, helping to simplifying your study.
Relevant Results for Your Electrophysiology Studies
Using Invitrogen™ B-27™ Electrophysiology Kit, allows you to get more relevant results when studying neuron physiology and the biochemical basis of normal and disease processes. The B-27™ Electrophysiology Kit can yield better results in other electrophysiological studies, including neurons used in multielectrode arrays in neurotoxicology and neuropharmacology studies.
For other applications, we offer alternative formulations of B-27™ and Neurobasal™.
Specifications
Specifications
| Content And Storage | B-27 Supplement, Electro (A14097-01) Store at -5°C to -20°C Neurobasal Media, Electro (A14098-01) Store at 2°C to 8°C |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Electrophysiology Kit |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| Product Line | Gibco |
| Quantity | 1 Kit |
Frequently Asked Questions (FAQs)
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?