missing translation for 'onlineSavingsMsg'
Learn More

Bollé Safety B-Clean Lens Cleaning Products/Kits

Product Code. 15382184 Shop All Bollé Safety Products
Change view
Click to view available options
Product Type:
Anti-Fog Spray
Cleaning Station
Lens Cleaner Spray
Multi-function Dry Cleaning Tissues
Tissue Dispenser
Quantity:
1 Ea.
100/Pk.
200/Pk.
250 mL
50 mL
500 mL
Unit Size:
250mL
500mL
50mL
Each
Pack of 100
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
15382184 Anti-Fog Spray 500 mL 500mL
12699090 Tissue Dispenser 100/Pk. Pack of 100
12659110 Cleaning Station 1 Ea. Each
12669110 Lens Cleaner Spray 250 mL 250mL
12659230 Multi-function Dry Cleaning Tissues 200/Pk. Pack of 200
10308429 Tissue Dispenser - Each
12649110 Lens Cleaner Spray 500 mL 500mL
12679110 Lens Cleaner Spray 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15382184 Supplier Bollé Safety Supplier No. PACF500
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

B-Clean Bollé Safety range of cleaning solutions guarantees optimum cleaning. Alcohol and silicone free, its water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles.

  • Alcohol and silicone free
  • Water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles
TRUSTED_SUSTAINABILITY

Specifications

Product Type Anti-Fog Spray
For Use With (Application) For All Types of Ocular
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • anti-fog kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1-methoxy-2-propanol, docusate sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.