missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ b-Cyclodextrin
Description
β-Cyclodextrin (β-CD) is a cyclic oligosaccharide comprising seven glucose subunits linked by α-1,4-glycosidic bonds, forming a cage-like supramolecular structure with a hydrophobic interior and a hydrophilic exterior. β-Cyclodextrins can form inclusion complexes with a diverse range of hydrophobic compounds. These complexes increase the aqueous solubility, dissolution rate, and bioavailability of the guest compounds.
Specifications
Specifications
| CAS | 7585-39-9 |
| Quantity | 1 g |
| Product Line | Tocris |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C42H70O35 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?