missing translation for 'onlineSavingsMsg'
Learn More

b-Tocopherol (1.0 mg/mL in hexane)(Racemic Mixture), TRC

Product Code. 30451615
Change view
Click to view available options
Quantity:
5x1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451615 5x1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451615 Supplier Toronto Research Chemicals Supplier No. TRCT5261735X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beta Tocopherol
CAS 148-03-8
Molecular Formula C28 H48 O2
Quantity 5x1 mL
Synonym 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyltridecyl)-, [2R*(4R*,8R*)]-, 6-Chromanol, 2,5,8-trimethyl-2-(4,8,12-trimethyltridecyl)- (8CI), rel-(2R)-3,4-Dihydro-2,5,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol, 2,5,8-Trimethyl-2-(4,8,12-trimethyltridecyl)-6-chromanol, 5,8-Dimethyltocol, Cumotocopherol, DL-β-Tocopherol, Neotocopherol, dl-β-Tocopherol, p-Xylotocopherol, β-Tocopherol
InChI Formula InChI=1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)cc(C)c2O1
IUPAC Name (2R)-2,5,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Molecular Weight (g/mol) 416.68
Formula Weight 416.3654
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.