missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ B02

Product Code. 18714099
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18714099 5 mg 5mg
18704099 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18714099 Supplier Tocris Bioscience™ Supplier No. 6392/5
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

RAD51 recombinase inhibitor

B02 is a RAD51 recombinase inhibitor, exhibiting selectivity for human RAD51 over the E.coli homologue RecA (IC50 values are 27.4 μM and >250 μM, respectively). Inhibits homologous recombination and increases cellular sensitivity to DNA damage in vitro. Enhances the therapeutic effect of cisplatin (Cat. No. 2251) on tumor cells in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1290541-46-6
Synonym 3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone
Quantity 5 mg
Molecular Formula C22H17N3O
Formula Weight 339.4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.