Learn More
B02, Tocris Bioscience™
RAD51 recombinase inhibitor
Brand: Tocris Bioscience 6392/5
This item is not returnable.
View return policy
Description
B02 is a RAD51 recombinase inhibitor, exhibiting selectivity for human RAD51 over the E.coli homologue RecA (IC50 values are 27.4 μM and >250 μM, respectively). Inhibits homologous recombination and increases cellular sensitivity to DNA damage in vitro. Enhances the therapeutic effect of cisplatin (Cat. No. 2251) on tumor cells in vivo.Specifications
| 1290541-46-6 | |
| 3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone | |
| 5 mg |