missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B30 Series Pre-Printed GHS CLP Chemical Labels

Product Code. 30041474 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
110.00 mm (W) x 75.00 mm (H)
84.99 mm (W) x 55.00 mm (H)
Display Text:
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
3 Red Diamonds per Sticker
Unit Size:
Pack of 270
Pack of 340
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Display Text Unit Size
30041474 110.00 mm (W) x 75.00 mm (H) 3 Red Diamonds per Sticker Pack of 270
30041405 110.00 mm (W) x 75.00 mm (H) 2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker Pack of 270
30041456 84.99 mm (W) x 55.00 mm (H) 3 Red Diamonds per Sticker Pack of 340
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041474 Supplier Brady™ Supplier No. 121642
  • ONLINE EXCLUSIVE PRICE
    273.00 €
    262.47 € / Pack of 270
    Save 10.53 €
    3% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-printed GHS/CLP chemical labels for BBP3X/S3XXX/i3300 printers.

For BBP3X/S3XXX/i3300 printers.

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and section 3
Description Pre-printed GHS/CLP Chemical Labels for BBP3X/S3XXX/i3300 Printers
Adhesive Type Permanent Acrylic
Coating None
Color Red on White
Finish Gloss
Material Vinyl
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 3 Red Diamonds per Sticker
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labelling, Safety Identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model B30-R10000
No. per Pack 270 Labels/Roll
Quantity 270 Stickers
Shape Rectangle
Size 110.00 mm (W) x 75.00 mm (H)
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 110.00 mm (W) x 75.00 mm (H)
Length (English) 2.953 in.
Length (Metric) 75.00 mm
Width (English) 4.331 in.
Width (Metric) 110.00 mm
Width (English) Printable Area 110.00 mm
Alternate Part Number B30-261-7569-CLP3B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.