missing translation for 'onlineSavingsMsg'
Learn More

Motic™ BA310POL Binocular Polarization Microscope

Product Code. 15485262
Change view
Click to view available options
Model:
BA310POL Binocular Polarization Microscope
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15485262 BA310POL Binocular Polarization Microscope Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485262 Supplier Motic™ Supplier No. 1100100402242
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use this polarization microscope for transmitted light samples. Motic™ BA310POL Binocular Microscope is designed for the examination of birefringent samples from petrography and mineralogy, and can also be used for the synthetic materials industry.

  • 30° inclined Binocular head for comfortable viewing while seated
  • Interpupillary distance adjustment between 55 to 75mm
  • Widefield high eyepoint, N-WF10X/20mm, diopter adjustment on both eyepieces, rubber eyecup (paired), cross hair on one eyepiece
  • Intermediate tube with rotatable analyzer 360° and Bertrand lens, focusable
  • Quartz wedge, full λ and 1/4λ retardation plates
  • Reversed quadruple nosepiece with click stops for precise magnification changes, individually centrable with one reference position
  • CCIS EC Plan Strain-free objectives EC PL 4X/0.10, 10X/0.25, 40X/0.65-Spring, 60X/0.80-Spring
  • Coaxial coarse and fine focusing system with 2 micron minimum increment with tension adjustment
  • Vertical travel range 20mm
  • 360° circular rotating stage with a diameter of 160mm, lockable position
  • Focusable and centrable Achromat swing-out condenser N.A. 0.90/0.13, Strain-free
  • Rotatable polarizer, fixed on condenser carrier
  • Collector lens assembly with screw-on filter holder
  • Koehler illumination quartz halogen 6V/30W with intensity control

Specifications

Condenser Type Achromat swing-out condenser N.A. 0.90/0.13 (strain-free) with iris diaphragm
Eyepiece Widefield High Eyepoint, N-WF10X/20 mm, diopter adjustment on both eyepieces, rubber eyecup (paired), cross hair on one eyepiece
Lighting Type Koehler Quartz Halogen with Intensity Control 6V/30w
Model BA310POL Binocular Polarization Microscope
Includes Quartz wedge, λ and 1/4λ retardation plates; Polarizing mechanical stage; C-mount camera adapters: 0.5X, 0.65X, 1X; Photo adapter, 2.5X photo eyepiece, 4X photo eyepiece
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.