missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bac-to-Bac™ C-His TOPO™ Cloning Kit

Product Code. 10014014
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10014014 20 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10014014 Supplier Invitrogen™ Supplier No. A11098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Bac-to-Bac™ C-His TOPO™ Cloning Kit

Bac-to-Bac™ Baculovirus Expression is an efficient method for producing baculovirus for high-level protein expression in insect cells. It relies on generation of recombinant virus by site-specific transposition in E. coli rather than homologous recombination in insect cells. New: pFastBacTOPO™ vectors!


Reliable and fast protein expression
The Bac-to-Bac™ Baculovirus Expression System is faster and easier than traditional baculovirus expression, and it maintains high levels of protein expression. Bac-to-Bac™ relies on generation of recombinant baculovirus by site-specific transposition in E. coli rather than homologous recombination in insect cells. The Bac-to-Bac™ Baculvirus Expression System is highly regarded in academic Literature. Approximately 100 citations every year since 2000!

Traditional baculovirus systems require purification and amplification of an initial low-titer viral supernatant. This requires a time-consuming plaque assay. Bac-to-Bac™'s pFastBac™ vector, however, recombines with the parent bacmid in DH10Bac™ E. coli competent cells to form an expression bacmid. Transfect the bacmid into insect cells for fast production of a high titer of pure recombinant baculovirus particles in the very first transfection. You'll save weeks of precious time. Collect a pure P2 baculovirus stock on Day 10 without the necessity of tedious plaques assay (See Figure 1)

High expression, easy screening
pFastBac™ uses the strong polyhedrin promoter to generate high levels of expression in a variety of insect cell lines such as Sf9, Sf21, and High Five™ cells.

New additions:
Bac-to-Bac™'s pFastBac vectors are now available as pFastBacTOPO™ vectors too. pFastBacTOPO™ vectors are Blunt™ TOPO™ vectors and are supplied with Mach1™-T1R E. coli for easy cloning and pFastBac⁄GOI propagation. The new vectors enable you to:

  • Amplify your gene of interest (GOI) with a PCR enzyme of highest fidelity such as Accuprime™ Pfx SuperMix.

  • TOPO™ clone the blunt-end PCR product into the new vector in only 5 minutes!

  • Visualize colonies 8 hours after plating on ampicillin selective plates because Mach1™-T1R E. coli cells have a faster doubling time compared to other standard cloning strains.

  • With the Bac-to-Bac™ C-His TOPO™ cloning or expression kit, you can produce a C-terminal His-fusion protein with a TEV cleavage site to purify with nickel-chelating resins (Invitrogen’s ProBond™ Purification System) and generate a native protein with the aid of Invitrogen’s popular AcTEV™ Protease. This is the Invitrogen’s only vector that contains a C-terminal TEV cleavage site!

    The new vectors combine the TOPO™ cloning technology with the highly regarded Bac-to-Bac™ baculovirus expression features to enable easy cloning and high-level protein expression.
    The pFastBacTOPO™ vectors are offered as cloning and expression kits.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Antibiotic Resistance Bacterial Ampicillin (AmpR), Gentamicin (GmR)
    Cell Line High Five™, Sf21, Sf9
    Cell Type Insect Cell
    Cloning Method Blunt TOPO
    Content And Storage Kit content for Bac-to-Bac™ C-His TOPO™ cloning kit (For details see the manual)
    • Vector kit for 20 topo cloning reactions:
    o pFastBac⁄CT-TOPO™ Vector containing the C-terminal TEV cleavage sit and His-Tag.
    o pFastBac⁄CT-Gus Control Plasmid (Control expression vector)
    o Other reagents supplied: 10x PCR, dNTP Mix, Salt Solution, Sterile Water, Control PCR template, Polyhedrin Forward Primer, SV40 pA Reverse Primer
    • Kit containing competent cells (20 reactions):
    o One Shot™ Mach1-T1R Chemically Competent E. coli

    Vector kit: store at -20°C
    One Shot™ Mach1-T1R Chemically Competent E. coli: store at -80°C
    Expression Mechanism Insect Cell Expression
    Expression System Baculovirus
    For Use With (Application) Protein Expression, Viral Vaccine Production
    No. of Reactions 20 Reactions
    Product Line Bac-to-Bac, TOPO
    Product Type Cloning Kit
    Promoter Polyhedrin
    Protein Tag His Tag (6x)
    Quantity 20 Reactions
    Selection Agent (Eukaryotic) Ampicillin, Gentamicin
    Vector pFastBac/CT-TOPO
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.