missing translation for 'onlineSavingsMsg'
Learn More

Hartmann Bacillol™ Zero Tissues

Product Code. 30048900
Change view
Click to view available options
Packaging Quantity:
Flow-Pack, 100 Tissues
Flow-Pack, 80 XL Tissues
Dimensions (L x W):
20 x 17 cm (7.9 x 6.7 in.)
20 x 30 cm (7.9 x 11.8 in.)
Size:
Standard
X-Large
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Dimensions (L x W) Size Unit Size
30048900 Flow-Pack, 100 Tissues 20 x 17 cm (7.9 x 6.7 in.) Standard Pack of 6
30058117 Flow-Pack, 80 XL Tissues 20 x 30 cm (7.9 x 11.8 in.) X-Large Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30048900 Supplier Hartmann Supplier No. 981935
  • ONLINE EXCLUSIVE PRICE
    94.89 €
    79.00 € / Pack of 6
    Save 15.89 €
    16% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleaning and disinfecting wipes based on organic acids, Bacillol™ Zero Tissues are highly effective and gentle on materials.

  • Ready-to-use quick disinfectant wipes made from 100% plastic-free material
  • Virucidal in 2 minutes
  • Free from alcohols, quaternary ammonium compounds, peroxides, perfumes, and dyes
  • >98.5% ingredients of organic origin
  • Patented formulation - organic acid complex
  • No hazardous substance labeling
  • Very good material compatibility
  • Sustainable, high-quality, non-woven wipe material
  • BPR-tested
  • Excellent, visible wetting of surfaces
  • Can be used without gloves
  • Good cleaning properties
  • Practical folding system for safe and easy removal of the individual wipes from the resealable flow-pack

Specifications

Product Type Disinfectant Wipe
Length (English) 7.9 in.
Length (Metric) 20 cm
Width (English) 6.7 in.
Width (Metric) 17 cm
Material Non-Woven Cloth
Autoclavable Not Autoclavable
Disposable Single-use
Dimensions (L x W) 20 x 17 cm (7.9 x 6.7 in.)
For Use With (Application) Cleaning and Disinfecting
Packaging Quantity Flow-Pack, 100 Tissues
Ply 1-ply
Size Standard
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.