Learn More
Thermo Scientific™ BacMam Akt [pSer473] Cellular Assay Kit
Description
The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT. BacMam provides a convenient genetic delivery tool for a GFP-AKT fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring phosphorylation of GFP-AKT at Ser473.
Specifications
Specifications
| Assay Entry | Cell-based terbium modification activity (transient), Cell-based terbium phospho-specific activity (transient) |
| Cell State | Cells Not Included |
| Content And Storage | Kit contains Terbium-labeled anti-AKT antibody, BacMam AKT Virus, and 6X Lysis Buffer. Store as indicated in handling documentation. |
| Description | BacMam Akt [pSer473] Cellular Assay Kit, 1 Kit, Fluorescent Detection Method, Kinases Druggable Target, TR-FRET Technique, LanthaScreen™ System, End Point Readout, GFP (EmGFP) Reporter |
| Detection Method | Fluorescence |
| For Use With (Application) | TR-FRET |
| Human Protein Class | Kinases, Cell Signalling |
| Includes | Terbium-labeled anti-AKT antibody, BacMam AKT virus and 6X lysis buffer |
| Product Line | LanthaScreen |
| Quantity | 1 kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.