missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BacMam Akt [pThr308] Cellular Assay Kit

Product Code. 10786041
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10786041

Brand: Thermo Scientific™ A12900

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT.

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT. BacMam provides a convenient genetic delivery tool for a GFP-AKT fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring phosphorylation of GFP-AKT at Thr308.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based terbium modification activity (transient), Cell-based terbium phospho-specific activity (transient)
Cell State Cells Not Included
Content And Storage Kit contains Terbium-labeled anti-AKT antibody, BacMam AKT Virus, and 6X Lysis Buffer. Store as indicated in handling documentation.
Detection Method Fluorescence
For Use With (Application) TR-FRET
Human Protein Class Kinases, Cell Signalling
Includes Cells not includes
Product Line LanthaScreen
Quantity 1 kit
Readout End Point
Reporter Gene GFP (EmGFP)
Target Entry AKT1
System Type LanthaScreen™
Druggable Target Kinases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.