Learn More
Thermo Scientific™ BacMam Histone H3 [AcLys9] Cellular Assay Kit
Description
The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of histones. BacMam provides a convenient genetic delivery tool for a GFP-Histone H3 fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring acetylation of GFP-Histone H3 at Lys9.
Specifications
Specifications
| Assay Entry | Cell-based terbium modification activity (transient) |
| Cell State | Cells Not Included |
| Content And Storage | Kit contains Terbium-labeled anti-Histone H3 antibody, BacMam Histone H3 Virus, and 6X Lysis Buffer. Store as indicated in handling documentation. |
| Detection Method | Fluorescence |
| For Use With (Application) | TR-FRET |
| Human Protein Class | Cell Signalling |
| Includes | Cells not includes |
| Product Line | LanthaScreen |
| Quantity | 1 Kit |
| Readout | End Point |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.