missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BacMam Histone H3K27me3 Cellular Assay Kit

Product Code. 11642149
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11642149

Brand: Invitrogen™ A14159

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The BacMam Histone H3K27me3 Cellular Assay is a high-throughput screening (HTS) compatible TR-FRET assay for the interrogation of trimethylation at lysine 27 on histone H3 in a cellular format.

The BacMam Histone H3K27me3 Cellular Assay is a high-throughput screening (HTS) compatible TR-FRET assay for the interrogation of trimethylation at lysine 27 on histone H3 in a cellular format. Baculovirus-mediated gene delivery (BacMam) provides a convenient tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest. Coupled with a terbium (Tb)-labeled site-specific antibody, the resulting cellular assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 27 of histone H3.

The kit includes:

  • BacMam Histone H3 Reagent
  • LanthaScreen™ Tb-anti-histone H3K27me3 Antibody
  • 6X LanthaScreen™ Cellular Lysis Buffer
  • Instrument Control Terbium TR-FRET kit

With the BacMam Histone H3K27me3 Cellular Assay Kit You Can:

  • Use your cell background of choice with the portability of BacMam
  • Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
  • Conserve precious cell samples with the miniaturizable homogenous assay format
  • Improve data quality with the advantages of TR-FRET

Get More Physiologically Relevant Results

The BacMam Histone H3K27me3 Cellular Assay allows the investigation of trimethylation at lysine 27 on histone H3 with your choice of cellular backgrounds including primary cells. This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys27 trimethylation in their natural complexes in a physiologically relevant cell type.

BacMam and LanthaScreen™ Convenience Saves Sample and Time

Assays are run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as Western blotting and ELISA, making them ideal for HTS applications. By using the same BacMam reagent for GFP-Histone H3, other histone H3 modifications (Ph, Me, Ac) can be monitored just be exchanging the Terbium (Tb) labeled antibody. In addition, the application of the LanthaScreen™ technology includes all the advantages of TR-FRET detection including reduced data noise, less interference from fluorescent compounds, and high sensitivity, allowing the use of fewer cells than traditional Western or ELISA methods.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based terbium modification activity (transient)
Cell State Cells Not Included
Content And Storage • BacMam Histone H3 Reagent
• LanthaScreen™ Tb-anti-histone H3K27me3 Antibody
• 6X LanthaScreen™ Cellular Lysis Buffer
• Instrument Control Terbium TR-FRET kit

Upon receipt:
Store the BacMam Histone H3 Reagent, 6X LanthaScreen™ Cellular Lysis Buffer, and Instrument Control Terbium TR-FRET kit at 4°C

Store the LanthaScreen™ Tb-anti-histone H3K27me3 Antibody at -20°C.
Description BacMam Histone H3K27me3 Cellular Assay Kit, 1 Kit, Fluorescent Detection Method, Demethylases, Methyltransferases Druggable Target, LanthaScreen™ System, 4 to –20°C Storage
Detection Method Fluorescence
For Use With (Application) TR-FRET
Includes BacMam Histone H3 Reagent, LanthaScreen™ Tb-anti-histone H3K27me3 Antibody, 6X LanthaScreen™ Cellular Lysis Buffer, instrument control terbium TR-FRET kit
Product Line LanthaScreen
Quantity 1 kit
Target Entry Histone H3
Type Cellular Assay Kit
System Type LanthaScreen™
Druggable Target Methyltransferases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.