Learn More
Cole-Parmer™ Coliform Test Kit
Product Code. 11737947 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
11737947 1 Each Each
1 options
Product Code. 11737947 Supplier Cole-Parmer™ Supplier No. 9956100
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Test kit screens for coliform bacterial contamination
  • Simply pour sample into bag, shake to dissolve dechlorinating tablet, pour into the five glass tubes, and store tubes at room temperature
  • Color change and small bubble formation provide indication of bacterial presence
Specifications
Includes Sterile water sampling bag with dechlorinating tablet, five 10mL glass tubes, and five coliform test tablets.
Quantity 1 Each
Type Coliform Test Kit